Learn More
Invitrogen™ HSPB11 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA5116808
Catalog No. PIPA5116808
Only null left
Add to Cart
Description
Description
Positive Samples: A-549, 293T, Mouse kidney, Mouse brain, Rat thymus Immunogen sequence: MRKIDLCLSS EGSEVILATS SDEKHPPENI IDGNPETFWT TTGMFPQEFI ICFHKHVRIE RLVIQSYFVQ TLKIEKSTSK EPVDFEQWIE KDLVHTEGQL QNEEIVAHDG SATYLRFIIV SAFDHFASVH SVSAEGTVVS NLSS.
HSPB11 (Heat shock protein beta-11), also known as IFT25, was originally identified as a small heat shock protein. Recently it has been identified as a component of IFT complex B. It has been demonstrated that IFT25 is not required for ciliary assembly but is required for proper Hedgehog signaling, which in mammals occurs within cilia. Cilia lacking IFT25 have defects in the signal-dependent transport of multiple Hedgehog components and fail to activate the pathway upon stimulation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HSPB11
Polyclonal
Unconjugated
Hspb11
2900042B11Rik; C1orf41; Heat shock protein; Heat shock protein beta-11; heat shock protein family B (small) member 11; heat shock protein family B (small), member 11; HSP; Hspb11; Hspb11 heat shock protein family B (small), member 11; HSPC034; HSPCO34; IFT25; intraflagellar transport 25 homolog; intraflagellar transport protein 25 homolog; Placental protein 25; placental protein 25 homolog; PP25
Rabbit
Affinity chromatography
RUO
51668, 685284, 72938
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.90 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q9D6H2, Q9Y547
Hspb11
Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human HSPB11 (NP_0572102).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.