Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspB2 Antibody (4B9E2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252490
Description
HspB2 Monoclonal specifically detects HspB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
HspB2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
DMPK-binding protein, HspB2, MKBP | |
Mouse | |
20.2 kDa | |
0.1 mg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
4B9E2 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
HSPB2 | |
Purified recombinant fragment of human HSPB2 (AA: 1-182) expressed in E. Coli. | |
Protein A or G purified | |
RUO | |
3316 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction