Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPB2 Monoclonal Antibody (4B9E2)
GREENER_CHOICE

Catalog No. PIMA531769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531769 Supplier Invitrogen™ Supplier No. MA531769
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31769 has been tested in indirect ELISA.

The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintenance of muscle structure and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4B9E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene HSPB2
Gene Accession No. Q16082
Gene Alias 27kDa; 2810021G24Rik; DMPK-binding protein; heat shock 27kD protein 2; heat shock 27kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein B2; heat shock protein beta 2; heat shock protein beta-2; heat shock protein family B (small) member 2; heat-shock protein beta-2; Hs.78846; HSP; HSP27; Hspb2; LOH11CR1K; MGC133245; MKBP
Gene Symbols HSPB2
Host Species Mouse
Immunogen Purified recombinant fragment of human HSPB2 (AA: 1-182) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3316
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.