Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579412
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579412 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579412 Supplier Invitrogen™ Supplier No. PA579412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Cardiac Muscle Tissue, Rat Skeletal Muscle Tissue.

The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintenance of muscle structure and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB2
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene HSPB2
Gene Accession No. O35878, Q16082
Gene Alias 27kDa; 2810021G24Rik; DMPK-binding protein; heat shock 27kD protein 2; heat shock 27kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein B2; heat shock protein beta 2; heat shock protein beta-2; heat shock protein family B (small) member 2; heat-shock protein beta-2; Hs.78846; HSP; HSP27; Hspb2; LOH11CR1K; MGC133245; MKBP
Gene Symbols HSPB2
Host Species Rabbit
Immunogen E.coli-derived human HSPB2 recombinant protein (Position: M1-P182).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161476, 3316
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.