Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106563 Supplier Thermo Scientific Supplier No. PA5106563
Add to Cart
Add to Cart

HSPB2 Polyclonal Antibody

HSPB2 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSPB2
Gene Accession No. O35878, Q16082, Q99PR8
Gene Alias 27kDa; 2810021G24Rik; DMPK-binding protein; heat shock 27kD protein 2; heat shock 27kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein B2; heat shock protein beta 2; heat shock protein beta-2; heat shock protein family B (small) member 2; heat-shock protein beta-2; Hs.78846; HSP; HSP27; Hspb2; LOH11CR1K; MGC133245; MKBP
Gene Symbols HSPB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSPB2(Accession Q16082), corresponding to amino acid residues L123-E173.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161476, 3316, 69253
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.