Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120807 Supplier Invitrogen™ Supplier No. PA5120807
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: BT-474, HeLa, Mouse skeletal muscle. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MSGRSVPHAH PATAEYEFAN PSRLGEQRFG EGLLPEEILT PTLYHGYYVR PRAAPAGEGS RAGASELRLS EGKFQAFLDV SHFTPDEVTV RTVDNLLEVS ARHPQRLDRH GFVSREFCRT YVLPADVDPW RVRAALSHDG ILNLEAPRGG RHLDTEVNEV YISLLPAPPD PEEEEEAAIV EP.

The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintenance of muscle structure and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSPB2
Gene Accession No. Q16082, Q99PR8
Gene Alias 27kDa; 2810021G24Rik; DMPK-binding protein; heat shock 27kD protein 2; heat shock 27kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein B2; heat shock protein beta 2; heat shock protein beta-2; heat shock protein family B (small) member 2; heat-shock protein beta-2; Hs.78846; HSP; HSP27; Hspb2; LOH11CR1K; MGC133245; MKBP
Gene Symbols HSPB2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-182 of human HSPB2 (NP_001532.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3316, 69253
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.