Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSPB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567826
Description
HSPB3 Polyclonal Antibody for Western Blot
Inhibitor of actin polymerization.
Specifications
HSPB3 | |
Polyclonal | |
Unconjugated | |
HSPB3 | |
DHMN2C; heat shock 17 kDa protein; heat shock 27kD protein 3; heat shock 27kD protein family, member 3; heat shock 27kDa protein 3; Heat shock protein; heat shock protein 3; heat shock protein B3; heat shock protein beta-3; heat shock protein family B (small) member 3; HMN2C; HSP; HSP 17; HSP27; HspB3; HSPL27; Protein 3 | |
Rabbit | |
affinity chromatography | |
RUO | |
8988 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q12988 | |
HSPB3 | |
Synthesized peptide derived from human HSPB3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction