Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100824 Supplier Invitrogen™ Supplier No. PA5100824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HSPB3.

Inhibitor of actin polymerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSPB3
Gene Accession No. Q12988, Q9QZ58
Gene Alias DHMN2C; heat shock 17 kDa protein; heat shock 27kD protein 3; heat shock 27kD protein family, member 3; heat shock 27kDa protein 3; Heat shock protein; heat shock protein 3; heat shock protein B3; heat shock protein beta-3; heat shock protein family B (small) member 3; HMN2C; HSP; HSP 17; HSP27; HspB3; HSPL27; Protein 3
Gene Symbols HSPB3
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSPB3(Accession Q12988), corresponding to amino acid residues T41-E91.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78951, 8988
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.