Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspB7 Antibody (OTI1D11), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271833UV
Description
HspB7 Monoclonal specifically detects HspB7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
HspB7 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HSPB7 | |
Full length recombinant protein of human HspB7 (NP_055239) produced in HEK293T cells. | |
0.1 mL | |
27129.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1D11 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Cardiovascular heat shock protein, CVHSP, DKFZp779D0968, FLJ32733, heat shock 27kDa protein family, member 7 (cardiovascular), heat shock protein beta-7, HspB7, member 7 (cardiovascular) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction