Learn More
Invitrogen™ HSPB8 Monoclonal Antibody (3C12-H11), APC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545253
Description
1 μg/mL of MA5-45253 was sufficient for detection of HSP22 in 20 μg of whole rat tissue extract by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 22kDa. Detects endogenous and exogenous HSP22 in monomeric, dimeric and tetrameric forms in WB. Does not cross react with alpha crystallin.
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease.
Specifications
HSPB8 | |
Monoclonal | |
1 mg/mL | |
2.15 mM MES, 47.35 mM phosphate with 1.033 mM EDTA and no preservative; pH 7.4 | |
Q9EPX0, Q9JK92, Q9UJY1 | |
HSPB8 | |
His-tagged human recombinant HSP22. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3C12-H11 | |
APC | |
HSPB8 | |
Alpha-crystallin C chain; AU018630; AW413033; CMT2L; Cryac; crystallin, alpha C; D5Ucla4; DHMN 2; DHMN2; E2IG1; E2-induced gene 1 protein; H11; H11K; heat shock 22kDa protein 8; heat shock 27kDa protein 8; Heat shock protein; heat shock protein 8; heat shock protein B8; heat shock protein beta-8; heat shock protein family B (small) member 8; HMN 2; HMN2; HMN2A; HSB8; HSP; HSP20-like; Hsp22; HSPB 8; Hspb8; OTTHUMP00000239768; PP1629; Protein kinase H11; small stress protein-like protein HSP22 | |
Mouse | |
Protein G | |
RUO | |
113906, 26353, 80888 | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.