Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPB8 Recombinant Rabbit Monoclonal Antibody (3K10W1)
GREENER_CHOICE

Catalog No. PIMA535177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535177 Supplier Invitrogen™ Supplier No. MA535177
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MADGQMPFSC HYPSRLRRDP FRDSPLSSRL LDDGFGMDPF PDDLTASWPD WALPRLSSAW PGTLRSGMVP RGPTATARFG VPAEGRTPPP FPGEPWKVCV.

The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3K10W1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSPB8
Gene Accession No. Q9EPX0, Q9JK92, Q9UJY1
Gene Alias Alpha-crystallin C chain; AU018630; AW413033; CMT2L; Cryac; crystallin, alpha C; D5Ucla4; DHMN 2; DHMN2; E2IG1; E2-induced gene 1 protein; H11; H11K; heat shock 22kDa protein 8; heat shock 27kDa protein 8; Heat shock protein; heat shock protein 8; heat shock protein B8; heat shock protein beta-8; heat shock protein family B (small) member 8; HMN 2; HMN2; HMN2A; HSB8; HSP; HSP20-like; Hsp22; HSPB 8; Hspb8; OTTHUMP00000239768; PP1629; Protein kinase H11; small stress protein-like protein HSP22
Gene Symbols HSPB8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HSPB8/HSP22 (Q9UJY1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113906, 26353, 80888
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.