Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPB9 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. FEPA5145459
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145459 200 μL
FEPA5145460 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145459 Supplier Invitrogen™ Supplier No. PA5145459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSPB9 gene ontology (GO) annotation includes protein binding and nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene HSPB9
Gene Accession No. Q9BQS6, Q9DAM3
Gene Alias 1700007H20Rik; Cancer/testis antigen 51; CT51; Heat shock protein; Heat shock protein beta-9; heat shock protein family B (small) member 9; heat shock protein, alpha-crystallin-related, B9; HSP; HSPB9; small heat shock protein B9
Gene Symbols HSPB9
Host Species Rabbit
Immunogen Recombinant Human HSPb9 protein, Met1-Thr158 (Accession #Q9BQS6).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75482, 94086
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.