Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspBAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180027
Description
HspBAP1 Polyclonal specifically detects HspBAP1 in Human samples. It is validated for Western Blot.Specifications
HspBAP1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ22623, heat shock 27 kDa associated protein, HSPB (heat shock 27kD) associated protein 1, HSPB (heat shock 27kDa) associated protein 1, HSPB1-associated protein 1,27 kDa heat shock protein-associated protein 1, PASS1FLJ39386, Protein associated with small stress protein 1, protein associating with small stress protein PASS1 | |
Rabbit | |
55 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_078886 | |
HSPBAP1 | |
Synthetic peptide directed towards the middle region of human HSPBAP1. Peptide sequence GCNLVFQVQGRKRWHLFPPEDTPFLYPTRIPYEESSVFSKINVVNPDLKR. | |
Affinity purified | |
RUO | |
79663 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction