Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPBAP1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HSPBAP1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5721
![]() ![]() |
Proteintech Group Inc
276311AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
HSPBAP1 (HSPB (heat shock 27kDa) associated protein 1), also known as PASS1 (protein associated with small stress protein 1), is a 488 amino acid cytoplasmic protein that contains one JMJC (Jumonji C) domain and shares 80% identity with its rat homolog, PASS1. Widely expressed with highest expression in ovary, thymus and pancreas, HSPBAP1 is thought to play a role in mediating cellular stress responses within the cell. Due to the presence of a JMJC domain, HSPBAP1 may be involved in chromatin remodeling events. Defects or translocations in the gene encoding HSPBAP1 are associated with renal cell carcinoma 1 (RCC1), suggesting a possible role for HSPBAP1 in carcinogenesis. Three isoforms of HSPBAP1 exist due to alternative splicing events.Specifications
HSPBAP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96EW2 | |
79663 | |
HSPBAP1 Fusion Protein Ag26428 | |
Primary | |
-20°C | |
HSPBAP1 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HSPB1 associated protein 1, HSPBAP1, PASS1 | |
HSPBAP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title