Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspBP1 Antibody (OTI1D5), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271761PE
Description
HspBP1 Monoclonal specifically detects HspBP1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.Specifications
HspBP1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
HSPBP1 | |
Full length recombinant protein of human HspBP1 (NP_036399) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG1 |
Flow Cytometry | |
OTI1D5 | |
Flow Cytometry | |
FES1, Heat shock protein-binding protein 1, Hsp70 binding protein 1, hsp70 interacting protein, hsp70-binding protein 1, Hsp70-binding protein 2, Hsp70-interacting protein 1, Hsp70-interacting protein 2, HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1, HSPBP, HspBP1, hspBP2 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
23640.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction