Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPBP1 Monoclonal Antibody (OTI1H4), TrueMAB™, OriGene
SDP

Catalog No. 502540981
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0981 30 μL
50-167-4243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0981 Supplier OriGene Supplier No. TA503317S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSPBP1 Monoclonal antibody specifically detects HSPBP1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen HSPBP1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1H4
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Lgr5
Gene Accession No. O75473, Q9Z1P4
Gene Alias 1500019G21Rik; FES1; Heat shock protein; heat shock protein-binding protein 1; HSP; hsp70 interacting protein; hsp70-binding protein 1; Hsp70-binding protein 2; hsp70-interacting protein; hsp70-interacting protein 1; Hsp70-interacting protein 2; HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1; HSPA (Hsp70) binding protein 1; HSPA binding protein, cytoplasmic cochaperone 1; Hspbp; HSPBP1; HspBP2; LOW QUALITY PROTEIN: hsp70-binding protein 1; MGC128574 protein; PP1845; SPA ) binding protein, cytoplasmic cochaperone 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 250-550 of human LGR5 produced in HEK293T Cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14160, 8549
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.