Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPBP1 Mouse anti-Human, Clone: OTI1F3, liquid, TrueMAB™
SDP

Catalog No. 501674313
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-4313 Supplier OriGene Supplier No. TA503412
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HspBP1 is an Hsp70-interacting protein that was first isolated from a human heart cDNA library using the yeast two-hybrid system (1,2). The ATPase domain of Hsp70 binds HspBP1 and upon binding, inhibits Hsp70 chaperone activity. More recently HspBP1 was found to promote nucleotide dissociation from Hsc70 (3) and is capable of altering the confirmation of the Hsp70 ATPase domain (4). HspBP1 is capable of regulating any Hsp70 activity that requires ATPase.

Specifications

Antigen HSPBP1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F3
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HSPBP1
Gene Accession No. Q9NZL4
Gene Alias FES1
Gene Symbols HSPBP1
Host Species Mouse
Immunogen Full length human recombit protein of human HSPBP1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23640
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.