Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPBP1 Polyclonal Antibody

Catalog No. PIPA5121892
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121892 20 μL
PIPA5119174 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121892 Supplier Invitrogen™ Supplier No. PA5121892
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSPBP1 Polyclonal Antibody for Western Blot, IHC (P)

HspBP1 is an Hsp70-interacting protein that was first isolated from a human heart cDNA library using the yeast two-hybrid system. The ATPase domain of Hsp70 binds HspBP1 and upon binding, inhibits Hsp70 chaperone activity. More recently HspBP1 was found to promote nucleotide dissociation from Hsc70 (3) and is capable of altering the confirmation of the Hsp70 ATPase domain. HspBP1 is capable of regulating any Hsp70 activity that requires ATPase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene HSPBP1
Gene Accession No. Q99P31, Q9NZL4
Gene Alias 1500019G21Rik; FES1; Heat shock protein; heat shock protein-binding protein 1; HSP; hsp70 interacting protein; hsp70-binding protein 1; Hsp70-binding protein 2; hsp70-interacting protein; hsp70-interacting protein 1; Hsp70-interacting protein 2; HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1; HSPA (Hsp70) binding protein 1; HSPA binding protein, cytoplasmic cochaperone 1; Hspbp; HSPBP1; HspBP2; LOW QUALITY PROTEIN: hsp70-binding protein 1; MGC128574 protein; PP1845; SPA ) binding protein, cytoplasmic cochaperone 1
Gene Symbols HSPBP1
Host Species Rabbit
Immunogen Recombinant protein Heat Shock 70kDa Binding Protein 1. The antigen corresponds to amino acid range 209-354 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100518376, 23640, 512757, 66245
Target Species Bovine, Human, Mouse, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.