Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPC132 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298141G
Description
HSPC132 Polyclonal specifically detects HSPC132 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HSPC132 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
15E1.1, MDM35, MDM-35, Mitochondrial Distribution And Morphology 35 Homolog, P53CSV, P53-Inducible Cell-Survival Factor, Protein 15E1.1, TP53 regulated inhibitor of apoptosis 1, TP53-Regulated Inhibitor Of Apoptosis, TRIAP1, WF-1 | |
Produced in rabbits immunized with E. coli-derived Human HSPC132 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
51499 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction