Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPC268 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | HSPC268 |
---|---|
Dilution | Western Blot 1:100-1:250, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HSPC268 Polyclonal specifically detects HSPC268 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HSPC268 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C7orf55, chromosome 7 open reading frame 55, FLJ35699, FMC1, formation of mitochondrial complexes 1 homolog, HSPC268 | |
C7ORF55 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 1:100-1:250, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q96HJ9 | |
154791 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MAALGSPSHTFRGLLRELRYLSAATGRPYRDTAAYRYLVKAFRAHRVTSEKLCRAQHE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title