Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89934316 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-316 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-316 Supplier Abnova Corporation Supplier No. PAB29783
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial synthetic peptide of rat Hspd1.

Sequence: YDAMLGDFVNMVEK

Specifications

Antigen HSPD1
Applications Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial synthetic peptide of rat Hspd1.
Dilution Immunocytochemistry (0.5-1 ug/mL) Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 5mg BSA, 0.9mg NaCl, 0.2mg Na
Gene Alias Hsp60
Gene Symbols Hspd1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 503-516 at C-terminus of rat Hspd1.
Purification Method Immunogen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63868
Test Specificity The synthetic peptide of this antibody is different from the related human sequence by one amino acid.
Target Species Human, Mouse, Rat
Content And Storage Store at −20°C. After reconstitution with 0.2mLof distilled water and concentration will be 500μg/mL, store at −20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.