Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA129794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA129794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA129794 Supplier Invitrogen™ Supplier No. PA129794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa Heat Shocked Cell LysateRecombinant Human Chaperonin 10 Protein.

The heat shock proteins (HSPs) comprise a group of highly conserved, abundantly expressed proteins with diverse functions, including the assembly and sequestering of multiprotein complexes, transportation of nascent polypeptide chains across cellular membranes and regulation of protein folding. Heat shock proteins (also known as molecular chaperones) fall into six general families: HSP 90, HSP 70, HSP 60, the low molecular weight HSPs, the immunophilins and the HSP 110 family. The low molecular weight family includes HSP 10, HSP 20, HSP 27 (Heme Oxygenase 1), HSP 32 and HSP 40. Chaperonins are ubiquitous, indispensable proteins that facilitate protein folding in an ATP-dependent manner, enhancing the yield of properly folded substrate protein under conditions where spontaneous folding does not occur. Chaperonins are typified by the E. coli heat-shock proteins GroEL and GroES (HSP 10). HSP 10 is a heptameric ring of identical 10 kDa subunits that binds to each end of GroEL to form a symmetric, functional heterodimer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPE1
Applications Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.2
Gene HSPE1
Gene Accession No. P26772, P61603, P61604, Q64433
Gene Alias 10 kDa chaperonin; 10 kDa heat shock protein, mitochondrial; 10 kDa heat shock protein, mitochondrial; MOB-like protein phocein; 10kDa; 10kDa Chaperonin; CH10; chaperonin 10; Cpn 10; CPN10; cpn10 protein; early pregnancy factor; early-pregnancy factor; EPF; GROES; Heat shock 10 kD protein 1 (chaperonin 10); heat shock 10 kDa protein 1 (chaperonin 10); heat shock 10 protein 1; heat shock 105kDa/110kDa protein 1; heat shock 10kD protein; heat shock 10kD protein 1 (chaperonin 10); heat shock 10kDa protein 1; heat shock 10kDa protein 1 (chaperonin 10); heat shock protein; heat shock protein 1 (chaperonin 10); heat shock protein 105; heat shock protein 105 kDa; heat shock protein 105 kDa; LOW QUALITY PROTEIN: heat shock protein 105 kDa; heat shock protein family E (Hsp10) member 1; heat shock protein family E (Hsp10) member 1 L homeolog; heat shock protein family H (Hsp110) member 1; HSP; hsp10; hsp105; hsp105a; hsp105b; hsp110; Hspe1; hspe1.L; Hsph1; hsph1-a; hsph1-b; mitochondrial 10 kDa heat shock protein; mitochondrial chaperonin 10; MOB family member 4, phocein; MOB1, Mps One Binder kinase activator-like 3; MOB4; MOBKL3; mob-like protein phocein; mt-cpn10; PREI3; preimplantation protein 3; XELAEV_18045050mg; zgc:86747
Gene Symbols HSPE1, hspe1.L, LOC100357698
Host Species Rabbit
Immunogen Synthetic peptide: RDGDILGKYVD conjugated to KLH, corresponding to amino acids 91-101 of Human Cpn10.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100350401, 100357698, 106024963, 15528, 25462, 281833, 3336, 394775, 397575, 414669, 478855
Target Species Bovine, Canine, Guinea Pig, Human, Mouse, Sheep, Pig, Rabbit, Rat, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.