Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSPE1 Recombinant Rabbit Monoclonal Antibody (JG82-34)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA534778
Description
Positive Control: Mouse liver tissue, rat testis tissue, human liver cancer tissue, human colon tissue, mouse kidney tissue.
The heat shock proteins (HSPs) comprise a group of highly conserved, abundantly expressed proteins with diverse functions, including the assembly and sequestering of multiprotein complexes, transportation of nascent polypeptide chains across cellular membranes and regulation of protein folding. Heat shock proteins (also known as molecular chaperones) fall into six general families: HSP 90, HSP 70, HSP 60, the low molecular weight HSPs, the immunophilins and the HSP 110 family. The low molecular weight family includes HSP 10, HSP 20, HSP 27 (Heme Oxygenase 1), HSP 32 and HSP 40. Chaperonins are ubiquitous, indispensable proteins that facilitate protein folding in an ATP-dependent manner, enhancing the yield of properly folded substrate protein under conditions where spontaneous folding does not occur. Chaperonins are typified by the E. coli heat-shock proteins GroEL and GroES (HSP 10). HSP 10 is a heptameric ring of identical 10 kDa subunits that binds to each end of GroEL to form a symmetric, functional heterodimer.
Specifications
HSPE1 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P26772, P61604, Q64433 | |
HSPE1 | |
Recombinant protein within Human Cpn10 aa 10-102. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
JG82-34 | |
Unconjugated | |
HSPE1 | |
10 kDa chaperonin; 10 kDa heat shock protein, mitochondrial; 10 kDa heat shock protein, mitochondrial; MOB-like protein phocein; 10kDa; 10kDa Chaperonin; CH10; chaperonin 10; Cpn 10; CPN10; cpn10 protein; early pregnancy factor; early-pregnancy factor; EPF; GROES; Heat shock 10 kD protein 1 (chaperonin 10); heat shock 10 kDa protein 1 (chaperonin 10); heat shock 10 protein 1; heat shock 105kDa/110kDa protein 1; heat shock 10kD protein; heat shock 10kD protein 1 (chaperonin 10); heat shock 10kDa protein 1; heat shock 10kDa protein 1 (chaperonin 10); heat shock protein; heat shock protein 1 (chaperonin 10); heat shock protein 105; heat shock protein 105 kDa; heat shock protein 105 kDa; LOW QUALITY PROTEIN: heat shock protein 105 kDa; heat shock protein family E (Hsp10) member 1; heat shock protein family E (Hsp10) member 1 L homeolog; heat shock protein family H (Hsp110) member 1; HSP; hsp10; hsp105; hsp105a; hsp105b; hsp110; Hspe1; hspe1.L; Hsph1; hsph1-a; hsph1-b; mitochondrial 10 kDa heat shock protein; mitochondrial chaperonin 10; MOB family member 4, phocein; MOB1, Mps One Binder kinase activator-like 3; MOB4; MOBKL3; mob-like protein phocein; mt-cpn10; PREI3; preimplantation protein 3; XELAEV_18045050mg; zgc:86747 | |
Rabbit | |
Protein A | |
RUO | |
15528, 25462, 3336 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction