Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPG2 Rat anti-Human, Mouse, Clone: SPM255, Abnova™

Catalog No. 89934321 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-321 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-321 Supplier Abnova Corporation Supplier No. MAB13086
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native mouse Hspg2.

Specifications

Antigen HSPG2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SPM255
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native mouse Hspg2.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS.
Gene Alias AI852380/Pcn/Plc/per
Gene Symbols Hspg2
Host Species Rat
Immunogen Murine EHS laminin preparation.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15530
Target Species Human, Mouse
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.