Learn More
Description
- Offer fast, accurate and precise molecular weight polymer determinations as well as increased sample throughput and greatly reduced solvent consumption and disposal costs.
- With the advent of today's higher performance solvent management systems, many polymers can now be analyzed accurately and reproducibly utilizing lower volume columns.
- Results in total analysis times of 7 minutes or less, thereby increasing sample throughput up to 6x while decreasing solvent usage from 35 to 50 mL to 5 mL or less per run.
Specifications
Specifications
| Diameter (Metric) | 6 mm |
| Length (Metric) | 150 mm |
| Particle Shape | Spherical |
| Particle Size | 4 μm |
| Quantity | 1/Ea. |
| USP Type | L38 |
| Product Type | Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.