Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation HSPgel HT MB-L Column

Catalog No. 50780752 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Solvents:
ODCB
THF
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Solvents
50-780-752 ODCB
50-780-740 THF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-780-752 Supplier Waters Corporation Supplier No. 186001781
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The new HSPgel columns are specifically designed for high speed GPC analysis.

  • They offer fast, accurate and precise molecular weight polymer determinations as well as increased sample throughput and greatly reduced solvent consumption and disposal costs.
  • With the advent of today's higher performance solvent management systems, many polymers can now be analyzed accurately and reproducibly utilizing lower volume columns.
  • This results in total analysis times of 7 minutes or less, thereby increasing sample throughput up to 6x while decreasing solvent usage from 35-50 mL to 5 mL or less per run.

Specifications

Diameter (Metric) 6 mm
Length (Metric) 150 mm
Particle Shape Spherical
Particle Size 5 μm
Quantity 1/Ea.
Solvents ODCB
USP Type L21
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.