Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPH1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728699 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8699 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8699 Supplier Proteintech Group Inc Supplier No. 133831AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes both HSP105alpha and HSP105beta isoforms. Western blot analysis using this antibody detected a major band around 100-110kD in Jurkat cells.

HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.

Specifications

Antigen HSPH1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSPH1
Gene Accession No. Q92598
Gene Alias Antigen NY CO 25, Heat shock 110 kDa protein, Heat shock protein 105 kDa, HSP105, HSP105A, HSP105B, HSP110, HSPH1, KIAA0201, NY CO 25
Gene Symbols HSPH1
Host Species Rabbit
Immunogen HSPH1 Fusion Protein Ag4224
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10808
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.