Description
- Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide an ACQUITY UPLC column with an HPLC column of equivalent selectivity and resolving power.
- Using the ACQUITY UPLC columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.
Specifications
Specifications
| Particle Shape | Spherical |
| Particle Size | 1.8 to 3.5 μm |
| Pore Size | 100 Å |
| Quantity | 2/Pk. |
| USP Type | L1 |
| Endcapped | No |
| Product Type | Method Transfer Kit, High Res |