Description
- ACQUITY UPLC HSS CN chemistry is an ultra-performance general purpose propyl cyano bonded phase that provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH.
Specifications
Specifications
| Particle Shape | Spherical |
| Particle Size | 1.8 to 3.5 μm |
| Pore Size | 100 Å |
| Quantity | 2/Pk. |
| USP Type | L10 |
| Endcapped | No |
| Product Type | Method Transfer Kit, High Res |