Learn More
Waters Corporation HSS PFP Method Transfer Kit
Catalog No. 50179446 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Particle Size:
1.8 to 3.5 μm
1.8 to 5 μm
Product Type:
Method Transfer Kit
Method Transfer Kit, High Res
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Product Type
50-179-446 1.8 to 3.5 μm Method Transfer Kit, High Res
50-179-444 1.8 to 5 μm Method Transfer Kit
50-179-445 1.8 to 3.5 μm Method Transfer Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-179-446 Supplier Waters Corporation Supplier No. 186005981
ACQUITY HSS PFP columns are fully scalable and transferable to HPLC separations.
  • ACQUITY UPLC HSS PFP is a non-endcapped silica-based column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase.
Specifications
Particle Shape Spherical
Particle Size 1.8 to 3.5 μm
Pore Size 100 Å
Quantity 1/Ea.
USP Type L43
Endcapped No
Product Type Method Transfer Kit, High Res