Learn More
Waters Corporation HSS T3 Method Transfer Kit
Catalog No. 50791280 Shop All Waters Corporation Products
Encompass_Preferred
Change view
Click to view available options
Particle Size:
1.8 to 3.5 μm
1.8 to 5 μm
Product Type:
Method Transfer Kit
Method Transfer Kit, High Res
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Product Type
50-791-280 1.8 to 3.5 μm Method Transfer Kit, High Res
50-791-273 1.8 to 3.5 μm Method Transfer Kit
50-791-266 1.8 to 5 μm Method Transfer Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-791-280 Supplier Waters Corporation Supplier No. 186004976
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms.
  • Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide an ACQUITY UPLC column with an HPLC column of equivalent selectivity and resolving power.
  • Using the ACQUITY UPLC columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.
Specifications
Particle Shape Spherical
Particle Size 1.8 to 3.5 μm
Pore Size 100 Å
Quantity 2/Pk.
USP Type L1
Endcapped Yes
Product Type Method Transfer Kit, High Res