Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HSV Tag Monoclonal Antibody,

Catalog No. 691713MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
40 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-171-3MI 40 μg
69-171-4MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-171-3MI Supplier MilliporeSigma™ Supplier No. 691713
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HSV Tag™ Antibody is a mouse monoclonal antibody (IgG1) that has high specificity and affinity for an 11-amino acid peptide derived from Herpes Simplex Virus (HSV) glycoprotein D. The peptide is encoded by the HSV Tag sequence in appropriate expression vectors and by the Positive Control 36mer in the Novagen NovaTope™ System. The purified antibody produces a strong signal at a 1:10,000 dilution on Western blots (Sakai 1996), dot blots, and colony lifts when used with the Novagen Anti-Mouse IgG AP Conjugate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSV Tag
Classification Monoclonal
Conjugate Unconjugated
Formulation Stabilized solution of 1mg/mL of pure antibody in 50% glycerol, includes positive control extract containing 31.1kDa Protein
Host Species Mouse
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity HSV tag peptide GlnProGluLeuAlaProGluAspProGluAsp , C-terminal or internal
Content And Storage -20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.