Learn More
Description
Specifications
Specifications
| Antigen | HSV Tag |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Formulation | Stabilized solution of 1mg/mL of pure antibody in 50% glycerol, includes positive control extract containing 31.1kDa Protein |
| Host Species | Mouse |
| Quantity | 200 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Test Specificity | HSV tag peptide GlnProGluLeuAlaProGluAspProGluAsp , C-terminal or internal |
| Content And Storage | -20°C |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.