Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSV Type 2 (strain G) Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA17492
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA17492 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA17492 Supplier Invitrogen™ Supplier No. PA17492
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

The PA1-7492 antibody reacts with Herpes Simplex virus 2. PA1-7492 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-7492 antibody was raised against HSV2 Strain G ICP + virion antigens.

Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSV Type 2 (strain G)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Herpes Simplex 2; Herpes Simplex II; herpes simplex virus 2; HHV-2; HSV Type 2; HSV Type II; HSV2; HSV-2
Host Species Sheep
Immunogen HSV2 Strain G ICP + virion antigens.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.