Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSV1 Monoclonal Antibody (10A3), Zeta

Catalog No. 502215941
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-221-5941 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-221-5941 Supplier ZETA CORPORATION Supplier No. Z2215MS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Cocktail

A recommended positive control tissue for this product is Skin with HSV infection, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.

HSV1 Monoclonal antibody specifically detects HSV1 in Human, Virus samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen HSV1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10A3
Concentration 200 μg/mL
Conjugate Unconjugated
Formulation tris with BSA, NP-40 and <0.1% sodium azide
Gene Alias Herpes; Herpes Simplex; Herpes Simplex 1; Herpes Simplex I; herpes simplex virus 1; Herpes Simplex Virus Type 1; Herpes Virus; HHV; HSV; HSV Type 1; HSV1; KITH_HHV1S
Host Species Mouse
Immunogen Detergent-solubilized herpes simplex virus (HSV) type 1 infected cells
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.