Learn More
Air-Tite™ HSW Soft-Ject™ Disposable Syringes
Catalog No. 1481753
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
20 mL
30 mL
50 mL
High-quality, three-part disposable rubber piston syringes
  • Lubricated synthetic rubber piston allows for smooth draw while positive stop helps prevent accidental pullout
  • Bold graduations are easily read and resist wipe off
  • ETO gas sterilized syringes, latex and pyrogen free
  • Packaged in individual sterile strip packs
  • Made in Germany
Certifications
FDA registered CE Mark, ISO 9001 certified.
Specifications
Certifications/Compliance FDA, CE, ISO 9001
Latex Free Yes
Product Type Soft-Ject™ Syringe
Disposable Single-use
Volume (Metric) 5 mL
Connection Type Luer Lock
Graduated Yes
Sterility Sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.