Learn More
R&D Systems™ PARP/Apoptosis Chemiluminescent Assay Kit
SDP
Catalog No. 4685096K Shop All Novus Products
Change view
Click to view available options
Quantity:
96 Tests
Catalog No. Quantity
46-850-96K 96 Tests
1 options
This item is not returnable. View return policy
Catalog No. 46-850-96K Supplier R&D Systems™ Supplier No. 4685096K
Add to Cart
Add to Cart
This item is not returnable. View return policy
R&D Systems™ HT Chemiluminescent PARP Apoptosis Assay Kit is an ELISA kit for measuring PARP activity in cell lysates before and during apoptosis.
  • Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
  • Do not use past expiration date.
  • Includes:
    • PBS Wash Solution
    • PBS + 0.1% Triton X-100 Wash Solution
    • 1X I-PAR Assay Buffer (contains 0.1mg/mL BSA)
    • PARP Substrate Cocktail
    • PARP Enzyme
    • Etoposide
    • Antibody Diluent
    • Anti-PAR Monoclonal Antibody
    • Goat Anti-Mouse-IgG-HRP Conjugate
    • PeroxyGlow™ A and B Chemiluminescent Substrates
    • Cell Extraction Buffer
  • Ideal for measuring PARP activity in cell extracts prepared before and during apoptosis.
  • Semi-quantitatively detects PAR deposited onto immobilized histone proteins in a 96-well format.
Specifications
Quantity 96 Tests
Content And Storage Store the unopened product at -20°C to -70°C
Product Line R&D Systems