Learn More
R&D Systems™ PARP/Apoptosis Colorimetric Assay Kit
Description
- Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
- Do not use past expiration date.
- Includes:
- PBS Wash Solution
- PBS + 0.1% Triton X-100 Wash Solution
- 1X I-PAR Assay Buffer (contains 0.1mg/mL BSA)
- PARP Substrate Cocktail
- PARP Enzyme
- Etoposide
- Antibody Diluent
- Anti-PAR Monoclonal Antibody
- Goat Anti-Mouse-IgG-HRP Conjugate
- TACS-Sapphire™
- Cell Extraction Buffer
- Ideal for measuring PARP activity in cell extracts prepared before and during apoptosis.
- Semi-quantitatively detects PAR deposited onto immobilized histone proteins in a 96-well format.
Specifications
Specifications
| Quantity | 96 Tests |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |