Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTF9C Antibody (OTI1C3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271830F
Description
HTF9C Monoclonal specifically detects HTF9C in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
HTF9C | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TRMT2A | |
Full length recombinant protein of human HTF9C (NP_073564) produced in HEK293T cells. | |
0.1 mL | |
27037.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1C3 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
HTF9CHpaII tiny fragments locus 9c protein, MGC102728, TRM2 tRNA methyltransferase 2 homolog A (S. cerevisiae), tRNA (uracil-5-)-methyltransferase homolog A, tRNA methyltransferase 2 homolog A | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction