Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTL Strefa DropSafe Medlance™ Plus Safety Lancets
Available in five different color-coded versions to meet specific blood sample size applications and address the variety of skin types
Supplier: HTL Strefa 7043
This item is not returnable.
View return policy
Description
Color coded to avoid mistakes.
Gentle
- Ultra-sharp needle optimally positioned during skin penetration
- Linear needle tracking eliminates vibrations to reduce pain and prevent tissue damage
- Low triggering force ensures gentle and comfortable contact with puncture site
- Ergonomically shaped safety lancet with reliable contact activation mechanism
- Does not require pre-loading, for very simple blood sampling
- Single-use, self-destructing device which cannot be reloaded
- Needle is concealed before and just after use, preventing sharps injuries
- Sterilized with 25 kGy gamma irradiation for safety
- “Technique-independent” design ensures adequate blood flow at minimum pain level
Cat. No. 22-192-001 - DropSafe Medlance Plus Light
- Perfect device for glucose level monitoring for people with normal skin
- Optimal penetration depth and needle width guarantee efficient blood sampling and high comfort
- 25ga needle helps to minimize side effects of blood sampling like pain or bruising after the test
- Assures larger blood samples, optimal/sufficient quantities for conducting most common capillary blood tests (glucose level, hemoglobin, cholesterol, blood-type studies, coagulation, blood gas, virology analyses and others)
- Quick incision, linear needle guiding and precise geometry guarantees right blood sample just after one single lancet
- Designed for capillary blood tests requiring larger blood samples
- Perfect device for performing analyses when patient's skin is tougher (men, physical workers)
- Combination of 21ga needle (0.8mm) and 2.4mm penetration depth assures over 10μL blood after one single lancet
- Cat. No. 22-192-004 - DropSafe Medlance Plus Special
- Perfect for heel sticks on neonates and finger sticks on adults
- Ultra-sharp blade ensures adequate blood volume and quick healing
Specifications
Sterile | |
25g | |
1.5mm | |
Safety Lancet | |
200 Pack |
Lite | |
Blood Glucose monitoring purpose | |
0.05 in. | |
Purple |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction