Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTLV-2 env 801 Monoclonal Antibody (8.A.033)
GREENER_CHOICE

Catalog No. PIMA191454
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA191454 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA191454 Supplier Invitrogen™ Supplier No. MA191454
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-91454 detects HTLV2 env 801 from human samples.

The HTLV-2 env 801 gene encodes the envelope glycoprotein, which is pivotal for virus entry into host cells. Human T-cell leukemia virus type 2 (HTLV-2) shares many structural features with HTLV-1, with the envelope (Env) protein being a critical component in mediating viral entry via interaction with cell surface receptors. The env gene product is synthesized as a precursor that is cleaved into the surface (SU) and transmembrane (TM) subunits. The SU subunit facilitates receptor binding, while the TM subunit aids in membrane fusion, enabling the virus to enter the host cell. HTLV-2 predominantly targets and transforms CD8+ T-cells, distinct from HTLV-1, which primarily transforms CD4 + T-cells. Understanding the structural and functional nuances of the HTLV-2 envelope protein is crucial as it presents a target for therapeutic interventions aimed at preventing viral entry and subsequent infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTLV-2 env 801
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8.A.033
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris HCl with no preservative; pH 7.5
Gene Alias HTLV; HTLV2; HTLV-II
Host Species Mouse
Immunogen Highly purified HTLV-II envelope 801 antigen.
Purification Method Protein G
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.