Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTN1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
| Antigen | HTN1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5122313
|
Invitrogen™
PA5122313 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5122312
![]() |
Invitrogen™
PA5122312 |
100 μL |
Each for $511.50
|
|
|||||
Description
This gene encodes a member of the histatin family of small, histidine-rich, cationic proteins. They function as antimicrobial peptides and are important components of the innate immune system. Histatins are found in saliva and exhibit antibacterial, antifungal activities and function in wound healing.Specifications
| HTN1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P15515 | |
| 3346 | |
| Asp20-Asn57 (Accession P15515), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| HTN1 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| HIS1; His1 12/38; His1 31/57; His1-(12-38)-peptide; His1-(31-57)-peptide; histatin 1; histatin-1; Histatin-2; Histidine-rich protein 1; HTN1; Post-PB protein; PPB | |
| HTN1 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title