Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HTR1A Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142775
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.
Specifications
HTR1A | |
Polyclonal | |
Unconjugated | |
HTR1A | |
5HT1A; 5-HT1A; 5-HT-1A; 5-HT1a receptor; 5-hydroxytryptamine (serotonin) receptor 1A; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; ADRB2RL1; ADRBRL1; G-21; Gpcr18; G-protein coupled receptor 18; guanine nucleotide-binding regulatory protein-coupled receptor; HTR1A; PFMCD; RAT5HT1A; Serotonin receptor 1A | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
15550, 3350 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P08908, Q64264 | |
HTR1A | |
Peptide with sequence C-ERKNERTAEAKRK (aa 322-334). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction