Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549346
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549346 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549346 Supplier Invitrogen™ Supplier No. PA549346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HTR1A Polyclonal Antibody for Western Blot, Flow

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR1A
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HTR1A
Gene Accession No. P08908
Gene Alias 5HT1A; 5-HT1A; 5-HT-1A; 5-HT1a receptor; 5-hydroxytryptamine (serotonin) receptor 1A; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; ADRB2RL1; ADRBRL1; G-21; Gpcr18; G-protein coupled receptor 18; guanine nucleotide-binding regulatory protein-coupled receptor; HTR1A; PFMCD; RAT5HT1A; Serotonin receptor 1A
Gene Symbols HTR1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 239-273 amino acids from the Central region of human HTR1A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3350
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.