Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR1B Polyclonal Antibody, Invitrogen™

Catalog No. PIPA565031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565031 Supplier Thermo Scientific Supplier No. PA565031

Rabbit Polyclonal Antibody

Immunogen sequence: MEEPGAQCAPP PPAGSETWVP QANLSSAPSQ NCSAKDYIYQ DS Highest antigen sequence identity to the following orthologs - mouse 67%, rat 72%.

Serotonin (5-hydroxytryptamine, 5-HT), originally discovered as a serum factor plays important roles in regulating diverse biological processes in central and peripheral nervous systems, cardiovascular systems, and gastrointestinal systems. Molecular cloning studies have identified 5 receptors belonging to the 5-HT1 subtypes. These receptors are negatively coupled to adenylyl cyclase. Radioligand binding studies suggest the presence of 5-HT1B (formerly known as 5-HT1DbR) sites in substantia nigra, dorsal subiculum, globus pallidus, and superior collicum. In situ hybridization studies show abundance of 5-HT1BR mRNA in CA1 pyramidal cells of hippocampus, Purkinje cells of cerebellum, olfactory tubercle, subthalamic nucleus and somewhat lower levels in the caudate-putamen, cortical areas, thalamic and hypothalamic nuclei.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR1B
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HTR1B
Gene Accession No. P28222
Gene Alias 5-HT<1B> receptor; 5ht1b; 5-HT1B; 5-HT-1B; 5HT1B Receptor; 5-HT1B receptor; 5-HT1B serotonin receptor; 5-HT1DB; 5-HT-1D-beta; 5-hydroxytryptamine (serotonin ) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled; 5-hydroxytryptamine receptor 1B; HTR1B; HTR1D2; HTR1DB; S12; serotonin (5-hydroxytryptamine (5HT)) receptor type 1B; serotonin 1D beta receptor; Serotonin receptor 1B
Gene Symbols HTR1B
Host Species Rabbit
Immunogen Recombinant Human HTR1B
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.