Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595901
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595901 Supplier Invitrogen™ Supplier No. PA595901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTAGHAATMI AIVWAISICI SIPPLFWRQA KAQEEMSDCL VNTSQISYTI YSTCGAFYIP SVLLIILYGR IYRAARNRIL NPPSLYGKRF TTAHLITGSA G; Positive Samples: U-251MG, SGC-7901, SH-SY5Y, Mouse brain, Mouse testis, Mouse stomach, Rat brain, Rat testis; Cellular Location: Cell membrane, Multi-pass membrane protein.

G-protein coupled receptor for serotonin; found in nociceptors throughout the body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR1D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HTR1D
Gene Accession No. P28221, P28565, Q61224
Gene Alias 5 - Hydroxytryptamine (serotonin) receptor 1D; 5HT1D; 5-HT1D; 5-HT-1D; 5HT1D Receptor; 5-HT1d receptor; 5-HT-1D-alpha; 5-HT1DR; 5-hydroxytryptamine (serotonin) receptor 1D; 5-hydroxytryptamine (serotonin) receptor 1D beta; 5-hydroxytryptamine (serotonin) receptor 1D, G protein-coupled; 5-hydroxytryptamine receptor 1D; AI853647; Gpcr14; G-protein coupled receptor 14; HT1d receptor; HT1DA; Htr1d; HTR1DA; Htr1db; HTRL; RDC4; RP23-32L5.3; serotonin 1D alpha receptor; Serotonin receptor 1D
Gene Symbols HTR1D
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 150-250 of human HTR1D (NP_0008551).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15552, 25323, 3352
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.