Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ HTR1E (Human) Recombinant Protein
Click to view available options
Quantity:
2 μg
Description
Human HTR1E full-length ORF (NP_000856.1) recombinant protein without tag.
Specifications
Specifications
For Use With (Application) | Antibody Production |
Gene ID (Entrez) | 3354 |
Name | 5-hydroxytryptamine (serotonin) receptor 1E |
Quantity | 2 μg |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Gene Alias | 5-HT1E |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction