Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR1F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106542 Supplier Invitrogen™ Supplier No. PA5106542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total 5HT1F Receptor.

5-HT1F is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. 5-HT1F is a member of the GPCR family. Expression of 5-HT1F has been reported in various brain structures, including cortex, hippocampus, trigeminal ganglia, and cerebral blood vessels. Expression in peripheral tissues is limited to uterus, mesentery, and artery. No expression has been detected in heart, kidney, liver, pancreas, or spleen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR1F
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HTR1F
Gene Accession No. P30939, P30940, Q02284
Gene Alias 5-HT-1E-beta; 5ht1f; 5-HT1F; 5-HT-1F; 5HT1F Receptor; 5-HT1f receptor; 5HT6; 5-hydroxytryptamine (serotonin) receptor 1E beta; 5-hydroxytryptamine (serotonin) receptor 1F; 5-hydroxytryptamine (serotonin) receptor 1F, G protein-coupled; 5-hydroxytryptamine receptor 1F; Htr1eb; HTR1EL; HTR1F; MR77; Serotonin receptor 1F; serotonin receptor gene (similar to Mm Htr1eb)
Gene Symbols HTR1F
Host Species Rabbit
Immunogen A synthesized peptide derived from human HTR1F(Accession P30939), corresponding to amino acid residues C106-M156.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15557, 3355, 60448
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.