Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HTR2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533302
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla (100%); Orangutan, Gibbon, Monkey (95%); Marmoset (85%). Antigen retrieval requires Proteinase K treatment.
In platelets and gut, serotonin plays a major role in cardiovascular function and motility of the gastrointestinal tract, respectively. Serotonin mediates its effects through several of G protein coupled receptors, designated 5-HT receptors or alternatively SR receptors. The SR-2 receptors are comprised of three subtypes, SR-2A, SR-2B and SR-2C, which activate phospholipase C and release intracellular stores of calcium in response to serotonin. SR-2A has a specific role in tracheal smooth muscle contraction, bronchoconstriction and mediating aldosterone production, and it is also thought to play a role in several psychiatric disorders, including depression and schizophrenia. SR-2B is expressed in embryonic and adult cardiovascular tissues, gut and brain and plays an important role in the pathology of cardiac disorders. SR-2C is thought to mediate the effects of atypical antipsychotic drugs.
Specifications
| HTR2A | |
| Polyclonal | |
| Unconjugated | |
| Htr2a | |
| 5-HT-2; 5Ht-2; 5-HT2 receptor; 5-HT2A; 5-HT-2A; 5HT2A Receptor; 5-HT2A receptor; 5-HT2A/2C; 5-HT2A/2C receptor; 5-hydroxytryptamine (serotonin) receptor 2A; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine 2 receptor; 5-hydroxytryptamine receptor 2A; E030013E04; Htr2; Htr-2; HTR2A; serotonin 5HT-2 receptor; serotonin 5-HT-2A receptor; serotonin receptor 2A; SR2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P28223 | |
| Htr2a | |
| Synthetic 20 amino acid peptide from C-terminal cytoplasmic domainhuman 5HT2A Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction