Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120747 Supplier Invitrogen™ Supplier No. PA5120747
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SH-SY5Y, Mouse brain. Immunogen sequence: MDILCEENTS LSSTTNSLMQ LNDDTRLYSN DFNSGEANTS DAFNWTVDSE NRTNLSCEGC LSPSCLSLLH LQEKNWSALL TAVVIILTIA GNILVIMAVS.

In platelets and gut, serotonin plays a major role in cardiovascular function and motility of the gastrointestinal tract, respectively. Serotonin mediates its effects through several of G protein coupled receptors, designated 5-HT receptors or alternatively SR receptors. The SR-2 receptors are comprised of three subtypes, SR-2A, SR-2B and SR-2C, which activate phospholipase C and release intracellular stores of calcium in response to serotonin. SR-2A has a specific role in tracheal smooth muscle contraction, bronchoconstriction and mediating aldosterone production, and it is also thought to play a role in several psychiatric disorders, including depression and schizophrenia. SR-2B is expressed in embryonic and adult cardiovascular tissues, gut and brain and plays an important role in the pathology of cardiac disorders. SR-2C is thought to mediate the effects of atypical antipsychotic drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR2A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Htr2a
Gene Accession No. P28223, P35363
Gene Alias 5-HT-2; 5Ht-2; 5-HT2 receptor; 5-HT2A; 5-HT-2A; 5HT2A Receptor; 5-HT2A receptor; 5-HT2A/2C; 5-HT2A/2C receptor; 5-hydroxytryptamine (serotonin) receptor 2A; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine 2 receptor; 5-hydroxytryptamine receptor 2A; E030013E04; Htr2; Htr-2; HTR2A; serotonin 5HT-2 receptor; serotonin 5-HT-2A receptor; serotonin receptor 2A; SR2A
Gene Symbols Htr2a
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15558, 3356
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.